Airfiltronix APF-2 Aluminum Mesh Pre-Filter, 1" Deep; 5/Box
Representative image only.
Airfiltronix APF-2 Aluminum Mesh Pre-Filter, 1" Deep; 5/Box
Airfiltronix | Mfr: APF-2| Item: 4401168
A filter for every application
₹76,700.00INR / Box of 5
₹76,700.00INR / Box of 5
Buy It Together
Current Item
Airfiltronix APF-2 Aluminum Mesh Pre-Filter, 1" Deep; 5/Box
Airfiltronix APF-2 Aluminum Mesh Pre-Filter, 1" Deep; 5/Box
4401168
₹76,700.00INR / Box of 5
+ + + =
Total Price 1451700.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ensure optimal performance of your ductless fume hood
  • Ideal condensation media for removing airborne greases and mists
  • Extends life of expensive carbon and HEPA filters by removing gross contaminants
More About this Item
Choose from a wide range of filters to remove pollutants. These are compatible with the Aitfiltronix range of ductless fume hoods.
Specifications
  • Compatible WithAirfiltronix ductless fume hoods 33705-16 to -22, ductless fume and particulate removal workstations 33705-02 to -08, portable fume/particle extractors 33705-43 to -46, and balance enclosure 44011-00
  • Length (InchShort)17
  • Width (InchShort)10
  • Depth (InchShort)1
  • TypeAluminum Mesh Pre-Filter, 1" Deep
  • DescriptionAluminum Mesh Pre-Filter, 1" Deep; 5/Box