WIKA Forged-Brass Liquid-Filled Gauge - Type 213.40
Accurately measure pressure in heavy industrial environments
  • NEMA 4X (IP65) rated
These high-quality gauges are intended for use in adverse service conditions where pulsation or vibration exists. The solid forged-brass case and socket is resistant to shock and vibration, and is suitable for hydraulics and compressors. Liquid fill is glycerine.
14,600.00 - ₹41,400.00INR / Each
25 variations of this product are available.
See All Product Options Below
25 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Dial Size (in) Max Pressure (PSI)
Availability Pricing
Wika 213.40 Forged Brass Industrial Gauge ~ 0 To 1000 PSI, 1/2 6805154 Mfr # 9314229
COMPARE
±1% full-scale (ANSI/ASME Grade B) 4 1000
₹37,900.00INR / Each
Wika 213.40 Forged Brass Industrial Gauge ~ 0 To 5000 PSI, 1/2 6805162 Mfr # 9314261
COMPARE
±1% full-scale (ANSI/ASME Grade B) 4 5000
₹41,400.00INR / Each
Wika 213.40 Forged Brass Industrial Gauge ~ 0 To 100 PSI, 2 1/2" 6843279 Mfr # 9310703
COMPARE
±2-1-2% full-scale 2.5 100
₹14,600.00INR / Each
Wika 213.40 Forged Brass Industrial Gauge ~ 0 To 160 PSI, 2 1/2" 6843280 Mfr # 9310711
COMPARE
±2-1-2% full-scale 2.5 160
₹14,600.00INR / Each
Wika 213.40 Forged Brass Industrial Gauge ~ 0 To 5000 PSI, 2 1/2" 6843281 Mfr # 9310819
COMPARE
±2-1-2% full-scale 2.5 5000
₹16,200.00INR / Each
Wika 213.40 Forged Brass Industrial Gauge ~ 0 To 60 PSI, 2 1/2" 6843282 Mfr # 9310690
COMPARE
±2-1-2% full-scale 2.5 60
₹14,600.00INR / Each
Wika 213.40 Forged Brass Industrial Gauge ~ 0 To 600 PSI, 2 1/2" 6843284 Mfr # 9310754
COMPARE
±2-1-2% full-scale 2.5 600
₹14,600.00INR / Each
Wika 213.40 Forged Brass Industrial Gauge ~ 0 To 1000 PSI, 2 1/2" 6843285 Mfr # 9310770
COMPARE
±2-1-2% full-scale 2.5 1000
₹14,600.00INR / Each
Wika 213.40 Forged Brass Industrial Gauge ~ 0 To 3000 PSI, 2 1/2" 6843286 Mfr # 9310800
COMPARE
±2-1-2% full-scale 2.5 3000
₹16,200.00INR / Each
Wika 9149872 GAUGE 2.5IN 30/100PSI/KPA 1/4 6843368 Mfr # 9149872
COMPARE
± 2-1-2% 2.5 100
₹16,000.00INR / Each
Wika 213.40 2.5 Forged Brass Industrial Gauge, 2.5" dial, 1/4"NPT(M), 0 to 1500 psi 6843373 Mfr # 9310789
COMPARE
±2.5% full-scale 2.5 1500
₹16,200.00INR / Each
Wika 9314652 GAUGE 4IN 30PSI 1/4 L 6843380 Mfr # 9314652
COMPARE
± 1% 4 30
₹37,900.00INR / Each
Wika 9314695 GAUGE 4IN 200PSI 1/4 L 6843382 Mfr # 9314695
COMPARE
4 200
₹37,900.00INR / Each
Wika 9318224 GAUGE 2.5IN 600PSI 1/4 B 6843391 Mfr # 9318224
COMPARE
±1% full scale 2.5 600
₹15,200.00INR / Each
Wika 9318275 GAUGE 2.5IN 3000PSI 1/4 B 6843393 Mfr # 9318275
COMPARE
±1% full scale 2.5 3000
₹16,800.00INR / Each
Wika 9310738 GAUGE 2.5IN 300PSI 1/4 L 6843467 Mfr # 9310738
COMPARE
±1% full scale 2.5 300
₹14,600.00INR / Each
Wika 9314660 GAUGE 4IN 60PSI 1/4 L 6843468 Mfr # 9314660
COMPARE
±2% full scale 4 60
₹37,900.00INR / Each
Wika 9318178 GAUGE 2.5IN 100PSI 1/4 B 6843472 Mfr # 9318178
COMPARE
±1% full scale 2.5 100
₹15,200.00INR / Each
Wika 213.40 Forged Brass Industrial Gauge, 30" Hg To 15 PSI; 1/4" NPT(M), 2.5" Dial 6855501 Mfr # 9318011
COMPARE
±2-1-2% full-scale 2.5 15
₹15,300.00INR / Each
Wika 213.40 Forged Brass Industrial Gauge, 30" Hg To 160 PSI; 1/4" NPT(M), 2.5" Dial 6855505 Mfr # 9318054
COMPARE
±2-1-2% full-scale 2.5 160
₹15,300.00INR / Each
Showing 1 - 20 of 25