Thermo Scientific Orion Standard Ag/AgCl pH Electrodes
Measure pH of virtually any sample
  • Available in a variety of styles to meet your measurement needs
Combination electrodes come with either a glass or epoxy body. Glass-body electrodes are ideals for laboratory use since they perform at higher temperatures and can be cleaned with common laboratory solvents. Epoxy-body electrodes are good for applications where rough handling and breakage are a problem. The electrodes have either refillable or sealed (gel-filled) references. All combination electrodes have a 3.3-ft (1-m) cable and BNC connector.
29,400.00 - ₹124,000.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Body Material Electrode Type Description
Availability Pricing
Thermo Scientific 9156DJWP pH Electode, Sealed, DJ, Epoxy; WP BNC WW-05711-58 Mfr # 9156DJWP
COMPARE
Epoxy Double junction pH Electode, Sealed/DJ/Epoxy; WP BNC
₹35,100.00INR / Each
Thermo Scientific 9109WP pH/ATC Triode, Sealed, DJ, Epoxy; EDIN, Banana WW-05711-76 Mfr # 9109WP
COMPARE
Epoxy Double junction pH/ATC Triode, Sealed/DJ/Epoxy; EDIN/Banana
₹124,000.00INR / Each
Thermo Scientific 9102BNWP pH Probe, Refillable, SJ, Glass; WP BNC WW-05712-06 Mfr # 9102BNWP
COMPARE
Glass Single junction pH Probe, Refillable/SJ/Glass; WP BNC
₹38,400.00INR / Each
Thermo Scientific 9103BNWP pH Probe, Refillable, SJ, Glass, Semi-Micro; WP BNC WW-05712-11 Mfr # 9103BNWP
COMPARE
Glass Single junction pH Probe, Refillable/SJ/Glass/Semi-Micro; WP BNC
₹46,200.00INR / Each
Thermo Scientific 9104BNWP pH Probe, Refillable, SJ, Glass, Rugged Bulb; WP BNC WW-05712-16 Mfr # 9104BNWP
COMPARE
Glass Single junction pH Probe, Refillable/SJ/Glass/Rugged Bulb; WP BNC
₹38,200.00INR / Each
Thermo Scientific 9162BNWP pH Probe, Refillable, SJ, Glass, Low-Resistance; WPBNC WW-05712-26 Mfr # 9162BNWP
COMPARE
Glass Single junction pH Probe, Refillable/SJ/Glass/Low-Resistance; WPBNC
₹48,700.00INR / Each
Thermo Scientific 9102DJWP pH Probe, Refillable, DJ, Glass; WP BNC WW-05712-32 Mfr # 9102DJWP
COMPARE
Glass Double junction pH Probe, Refillable/DJ/Glass; WP BNC
₹38,400.00INR / Each
Thermo Scientific 9110DJWP pH Probe, Refillable, DJ, Glass, Semi-Micro; WP BNC WW-05712-34 Mfr # 9110DJWP
COMPARE
Glass Double junction pH Probe, Refillable/DJ/Glass/Semi-Micro; WP BNC
₹43,700.00INR / Each
Thermo Scientific 9156BNWP pH Probe, Refillable, SJ, Epoxy; WP BNC WW-05712-71 Mfr # 9156BNWP
COMPARE
Epoxy Single junction pH Probe, Refillable/SJ/Epoxy; WP BNC
₹29,400.00INR / Each
Thermo Scientific 9172BNWP Sure-Flow pH Probe, Refillable, SJ, Glass; WP BNC WW-05718-55 Mfr # 9172BNWP
COMPARE
Glass Single junction Sure-Flow pH Probe, Refillable/SJ/Glass; WP BNC
₹43,200.00INR / Each
Thermo Scientific 9165BNWP Sure-Flow pH Probe, Refillable, SJ, Epoxy; WP BNC WW-05718-57 Mfr # 9165BNWP
COMPARE
Epoxy Single junction Sure-Flow pH Probe, Refillable/SJ/Epoxy; WP BNC
₹43,000.00INR / Each
Thermo Scientific 9157BNMD pH/ATC Probe, Refillable, SJ, Epoxy; BNC/8-Pin DIN WW-05719-14 Mfr # 9157BNMD
COMPARE
Epoxy Single junction pH/ATC Probe, Refillable/SJ/Epoxy; BNC/8-Pin DIN
₹49,300.00INR / Each
Thermo Scientific 9107BN pH/ATC Probe; Sealed, SJ, Epoxy, BNC/8-pin DIN WW-58819-91 Mfr # 9107BN
COMPARE
Epoxy Single junction pH/ATC Probe; Sealed/SJ/Epoxy, BNC/8-pin DIN
₹37,300.00INR / Each
Thermo Scientific 9107WMMD pH/ATC Probe, Sealed, SJ, Epoxy, 10 ft; BNC/8-Pin DIN WW-58822-30 Mfr # 9107WMMD
COMPARE
Epoxy Single junction pH/ATC Probe, Sealed/SJ/Epoxy/10 ft; BNC/8-Pin DIN
₹39,800.00INR / Each
Thermo Scientific 9107WLMD pH/ATC Triode, epoxy body, sealed, single-junction, built-in ATC with 8 pin miniDIN connectio WW-58822-31 Mfr # 9107WLMD
COMPARE
Epoxy pH/ATC Triode, epoxy body, sealed, single-junction, built-in ATC with 8 pin miniDIN connectio
₹46,700.00INR / Each
Thermo Scientific 9107BNMD pH/ATC Probe, Sealed, SJ, Epoxy, 5 ft; BNC/8-Pin DIN WW-58823-30 Mfr # 9107BNMD
COMPARE
Epoxy Single junction pH/ATC Probe, Sealed/SJ/Epoxy/5 ft; BNC/8-Pin DIN
₹36,400.00INR / Each
Showing 1- 16 of 16