Thermo Scientific 9157BNMD pH/ATC Probe, Refillable, SJ, Epoxy; BNC/8-Pin DIN
Representative image only.
Thermo Scientific 9157BNMD pH/ATC Probe, Refillable, SJ, Epoxy; BNC/8-Pin DIN
Thermo Scientific | Mfr: 9157BNMD| Item: 0571914
Measure pH of virtually any sample
₹68,900.00INR / Each
₹68,900.00INR / Each
Buy It Together
Current Item
Thermo Scientific 9157BNMD pH/ATC Probe, Refillable, SJ, Epoxy; BNC/8-Pin DIN
Thermo Scientific 9157BNMD pH/ATC Probe, Refillable, SJ, Epoxy; BNC/8-Pin DIN
0571914
₹68,900.00INR / Each
+ =
Total Price 164520.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Available in a variety of styles to meet your measurement needs
  • 1 year warranty
More About this Item
Combination electrodes come with either a glass or epoxy body. Glass-body electrodes are ideals for laboratory use since they perform at higher temperatures and can be cleaned with common laboratory solvents. Epoxy-body electrodes are good for applications where rough handling and breakage are a problem. The electrodes have either refillable or sealed (gel-filled) references. All combination electrodes have a 3.3-ft (1-m) cable and BNC connector.
1 Year Warranty
Specifications
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionGlass fiber
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)90
  • Temperature CompensationYes
  • ATC Connector8-pin mini-DIN
  • ConnectorBNC
  • Cable Length (m)1
  • DescriptionpH/ATC Probe, Refillable/SJ/Epoxy; BNC/8-Pin DIN