Thermo Scientific Orion Star A100 Series Portable pH/mV/Temperature Meter
Combine simplicity with accuracy for your portable pH measurements
  • Large LCD displays pH or mV along with temperature
  • Up to 3 point calibration, with easy recall of slope data
  • Auto-Read™ function locks in stable reading
  • Automatic recognition of USA/NIST and DIN buffers
  • Nonvolatile memory holds up to 50 data points
  • Waterproof, IP67-rated housing
  • Four AA batteries provide over 2000 hours of field use, or order optional AC adapter for lab use
Thermo Scientific Orion Star A121 portable pH meter is ideal for field testing--it features waterproof and durable IP67-rated housing. Large informative display shows pH and mV readings with temperature, battery life, electrode status, meter mode, calibration data, power source, and more. Simple button layout and on-screen messages help with calibration and setup menu choices.
2 PRODUCT OPTIONS
Compareitem specifications
Max mV (mV) Max pH (pH) Description
Availability Pricing
Thermo Scientific STAR A121 Star A121 pH Portable Meter Only 5882506 Mfr # STARA1210
COMPARE
1999 16 Star A121 pH Portable Meter Only
₹95,620.00INR / Each
Thermo Scientific STAR A121 Star A121 pH Portable Meter Kit 5882508 Mfr # STARA1215
COMPARE
1999 16 Star A121 pH Portable Meter Kit
₹164,440.00INR / Each