Thermo Scientific 9103BNWP pH Probe, Refillable, SJ, Glass, Semi-Micro; WP BNC
Representative image only.
Thermo Scientific 9103BNWP pH Probe, Refillable, SJ, Glass, Semi-Micro; WP BNC
Thermo Scientific | Mfr: 9103BNWP| Item: 0571211
Measure pH of virtually any sample
₹65,700.00INR / Each
₹65,700.00INR / Each
Buy It Together
Current Item
Thermo Scientific 9103BNWP pH Probe, Refillable, SJ, Glass, Semi-Micro; WP BNC
Thermo Scientific 9103BNWP pH Probe, Refillable, SJ, Glass, Semi-Micro; WP BNC
0571211
₹65,700.00INR / Each
+ + =
Total Price 189120.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Available in a variety of styles to meet your measurement needs
  • 1 year warranty
More About this Item
Combination electrodes come with either a glass or epoxy body. Glass-body electrodes are ideals for laboratory use since they perform at higher temperatures and can be cleaned with common laboratory solvents. Epoxy-body electrodes are good for applications where rough handling and breakage are a problem. The electrodes have either refillable or sealed (gel-filled) references. All combination electrodes have a 3.3-ft (1-m) cable and BNC connector.
Glass-body, refillable, combination, semi-micro pH electrode for accurate test tube use with as little as 0.2 to 0.4 mL volume.
1 Year Warranty
Specifications
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)90
  • ConnectorBNC (waterproof when used with Thermo Scientific Orion Star Series meters)
  • Cable Length (m)1
  • DescriptionpH Probe, Refillable/SJ/Glass/Semi-Micro; WP BNC