Thermo Scientific 9165BNWP Sure-Flow pH Probe, Refillable, SJ, Epoxy; WP BNC
Representative image only.
Thermo Scientific 9165BNWP Sure-Flow pH Probe, Refillable, SJ, Epoxy; WP BNC
Thermo Scientific | Mfr: 9165BNWP| Item: 0571857
Measure pH of virtually any sample
₹61,210.00INR / Each
₹61,210.00INR / Each
Buy It Together
Current Item
Thermo Scientific 9165BNWP Sure-Flow pH Probe, Refillable, SJ, Epoxy; WP BNC
Thermo Scientific 9165BNWP Sure-Flow pH Probe, Refillable, SJ, Epoxy; WP BNC
0571857
₹61,210.00INR / Each
+ + =
Total Price 184630.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Available in a variety of styles to meet your measurement needs
  • 1 year warranty
More About this Item
Combination electrodes come with either a glass or epoxy body. Glass-body electrodes are ideals for laboratory use since they perform at higher temperatures and can be cleaned with common laboratory solvents. Epoxy-body electrodes are good for applications where rough handling and breakage are a problem. The electrodes have either refillable or sealed (gel-filled) references. All combination electrodes have a 3.3-ft (1-m) cable and BNC connector.
1 Year Warranty
Specifications
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionSure-flow
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • ConnectorBNC (waterproof when used with Thermo Scientific Orion Star Series meters)
  • Cable Length (m)1
  • DescriptionSure-Flow pH Probe, Refillable/SJ/Epoxy; WP BNC