Thermo Scientific 9156BNWP pH Probe, Refillable, SJ, Epoxy; WP BNC
Representative image only.
Thermo Scientific 9156BNWP pH Probe, Refillable, SJ, Epoxy; WP BNC
Thermo Scientific | Mfr: 9156BNWP| Item: 0571271
Measure pH of virtually any sample
₹41,880.00INR / Each
₹41,880.00INR / Each
Buy It Together
Current Item
Thermo Scientific 9156BNWP pH Probe, Refillable, SJ, Epoxy; WP BNC
Thermo Scientific 9156BNWP pH Probe, Refillable, SJ, Epoxy; WP BNC
0571271
₹41,880.00INR / Each
+ =
Total Price 137500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Available in a variety of styles to meet your measurement needs
  • 1 year warranty
More About this Item
Combination electrodes come with either a glass or epoxy body. Glass-body electrodes are ideals for laboratory use since they perform at higher temperatures and can be cleaned with common laboratory solvents. Epoxy-body electrodes are good for applications where rough handling and breakage are a problem. The electrodes have either refillable or sealed (gel-filled) references. All combination electrodes have a 3.3-ft (1-m) cable and BNC connector.
Epoxy-body, refillable, combination pH electrode with bulb guard is perfect for rugged application in the lab or out in the field.
1 Year Warranty
Specifications
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionGlass fiber
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)90
  • ConnectorBNC (waterproof when used with Thermo Scientific Orion Star Series meters)
  • Cable Length (m)1
  • DescriptionpH Probe, Refillable/SJ/Epoxy; WP BNC