Poxygrid High-Capacity Test Tube Racks, Epoxy-coated steel
Organize and process high volumes of samples
Epoxy-coated steel wire racks are chemical and corrosion resistant; unaffected by organic solvents, salts, weak acids, and weak alkalis. Epoxy will withstand a temperature range of -369 to 250°F (-223 to 121°C). Autoclavable
9,300.00 - ₹11,200.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Tube Size (mm) Tubes Held Description
Availability Pricing
Poxygrid F18751-0160 60 Place Epoxy-Coated Steel Rack for 10-13 mm Test Tubes; 1/Pk WW-06739-31 Mfr # F18751-0160
COMPARE
13 60 60 Place Epoxy-Coated Steel Rack for 10-13 mm Test Tubes; 1/Pk
₹9,300.00INR / Each
Poxygrid F18752-1150 Wire Test Tube Rack, 150, 10-13mm WW-06739-32 Mfr # F18752-1150
COMPARE
13 150 Wire Test Tube Rack, 150, 10-13mm
₹11,200.00INR / Each
Poxygrid F18756-0160 Wire Test Tube Rack, 60, 15-16mm WW-06739-33 Mfr # F18756-0160
COMPARE
16 60 Wire Test Tube Rack, 60, 15-16mm
₹9,900.00INR / Each
Poxygrid F18760-1150 Wire Test Tube Rack, 150, 15-16mm WW-06739-34 Mfr # F18760-1150
COMPARE
16 150 Wire Test Tube Rack, 150, 15-16mm
₹11,100.00INR / Each
Poxygrid F18763-0160 Wire Test Tube Rack, 60, 18-20mm WW-06739-35 Mfr # F18763-0160
COMPARE
20 60 Wire Test Tube Rack, 60, 18-20mm
₹10,600.00INR / Each
Poxygrid F18774-0160 Wire Test Tube Rack, 60, 22-25mm WW-06739-36 Mfr # F18774-0160
COMPARE
25 60 Wire Test Tube Rack, 60, 22-25mm
₹10,700.00INR / Each