In Stock
Width (in)
Height (in)
Length (in)
Max Tube Size (mm)
Tubes Held
Material
Industry
Poxygrid
Poxygrid
Scienceware® Poxygrid racks are coated with heat-cured epoxy, which is unaffected by organic solvents, salts, weak acids, and weak alkalis, for long life and easy cleaning. Choose test tube racks and drain stands. We are a Poxygrid authorized distributor.