Poxygrid Test Tube Half Racks, Epoxy-coated steel
Use when space is limited
Epoxy-coated steel wire racks are chemical and corrosion resistant; unaffected by organic solvents, salts, weak acids and alkalies. Epoxy will withstand a temperature range of -369 to 250°F (-223 to 121°C). Autoclavable.
8,500.00 - ₹8,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Tube Size (mm) Tubes Held
Availability Pricing
Poxygrid F18788-1300 Wire Test Tube Half Rack 36 10-13mm WW-06739-10 Mfr # F18788-1300
COMPARE
13 36
₹8,700.00INR / Each
Poxygrid F18788-1600 Wire Test Tube Half Rack 24 15-16mm WW-06739-12 Mfr # F18788-1600
COMPARE
16 24
₹8,500.00INR / Each
Poxygrid F18788-2000 Wire Test Tube Half Rack 20 18-20mm WW-06739-14 Mfr # F18788-2000
COMPARE
20 20
₹8,500.00INR / Each