Motic® Moticam Microscope Cameras
Capture images and stream video from your Motic® microscopes
  • Documentation and analysis for a deeper view of your slide
  • Easy to install and ready for immediate use
  • Capture, annotate, measure or evaluate still images and video
Motic® Moticam microscope cameras are designed for ease of use and the ability to output images and video. These cameras can produce still image and video capture, still image measurement, image adjustments, white balance, automatic and manual exposure, and individual objective calibration system.
Cameras are available in a wide range of resolution and connectivity options for applications ranging from education, conservation, pharmaceutical, or other demanding fields.
108,000.00 - ₹289,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Data Transfer Image Sensor Resolution
Availability Pricing
Motic Instruments Moticam A5 Microscope Camera, sCMOS Sensor, 5MP, 2592 x 1944 pixels, USB 4892593 Mfr # D-MOTICAM A5
COMPARE
USB 1/2.8" sCMOS 5MP
₹122,000.00INR / Each
Motic Instruments Moticam A8 Microscope Camera, sCMOS Sensor, 8MP, 3840 x 2160 pixels, USB 4892594 Mfr # 1100600101443
COMPARE
USB 1/3" sCMOS 8MP
₹130,000.00INR / Each
Motic Instruments Moticam A16 Microscope Camera, sCMOS Sensor, 16MP, 4608 x 3456 pixels, USB 4892595 Mfr # 1100600101463
COMPARE
USB 1/2.3" sCMOS 16MP
₹161,000.00INR / Each
Motic Instruments Moticam 4000 Microscope Camera, IMX334 Sensor, 8MP, 3840 x 2160 pixels, USB and HDMI 4892596 Mfr # 1100600101521
COMPARE
USB and HDMI 1/1.8" IMX334 8MP
₹289,000.00INR / Each
Motic Instruments Moticam X5 Microscope Camera, CMOS Sensor, 4MP, 1920 x 1080 pixels, Wi-Fi 4892598 Mfr # D-MOTICAM X5
COMPARE
Wi-Fi (2.4 & 5 GHz), Ethernet 1/3" CMOS 4MP
₹108,000.00INR / Each