Motic Instruments Moticam A5 Microscope Camera, sCMOS Sensor, 5MP, 2592 x 1944 pixels, USB
Representative image only.
Motic Instruments Moticam A5 Microscope Camera, sCMOS Sensor, 5MP, 2592 x 1944 pixels, USB
Motic Instruments | Mfr: D-MOTICAM A5| Item: 4892593
Capture images and stream video from your Motic® microscopes
₹122,000.00INR / Each
₹122,000.00INR / Each
Product Description
Includes
CS ring adapter, focusable lens, 30mm and 38mm eyepiece adapters, USB 2.0 cable, Motic 4-dot calibration slide, macro tube, and calibration card.
Key Features
  • Documentation and analysis for a deeper view of your slide
  • Easy to install and ready for immediate use
  • Capture, annotate, measure or evaluate still images and video
  • Moticam A5 features a 1/2.8" sensor size
  • 1 year warranty
More About this Item
Motic® Moticam microscope cameras are designed for ease of use and the ability to output images and video. These cameras can produce still image and video capture, still image measurement, image adjustments, white balance, automatic and manual exposure, and individual objective calibration system.
Cameras are available in a wide range of resolution and connectivity options for applications ranging from education, conservation, pharmaceutical, or other demanding fields.
At full 5MP live resolution, the Moticam A5 delivers up to 30 frames per second at 2592x1944 pixels per frame. Given its higher pixel count as well as sensitive sCMOS construction, the Moticam A5 performs well in many different contrasting techniques.
1 Year WarrantyConformite Europeenne
Specifications
  • Resolution5MP
  • Image Sensor1/2.8" sCMOS
  • Data Format(s)Still image JPEG/TIFF; Video MP4
  • System RequirementsMicrosoft Windows 10, Mac OSX10.9, Linux UBUNTU1604 or higher
  • Data TransferUSB
  • Power5V (USB supply)
  • DescriptionMicroscope Camera, sCMOS Sensor, 5MP, 2592 x 1944 pixels, USB
  • Warranty1 year warranty