Motic Instruments Moticam 4000 Microscope Camera, IMX334 Sensor, 8MP, 3840 x 2160 pixels, USB and HDMI
Representative image only.
Motic Instruments Moticam 4000 Microscope Camera, IMX334 Sensor, 8MP, 3840 x 2160 pixels, USB and HDMI
Motic Instruments | Mfr: 1100600101521| Item: 4892596
Capture images and stream video from your Motic® microscopes
₹289,000.00INR / Each
₹289,000.00INR / Each
Product Description
Includes
12 V power adapter, CS ring, Motic 4-dot calibration slide, HDMI cable, 12 mm lens, 30 mm to 38 mm adapter, and macro tube.
Key Features
  • Documentation and analysis for a deeper view of your slide
  • Easy to install and ready for immediate use
  • Capture, annotate, measure or evaluate still images and video
  • Moticam 4000 features a 1/1.8" sensor size
More About this Item
Motic® Moticam microscope cameras are designed for ease of use and the ability to output images and video. These cameras can produce still image and video capture, still image measurement, image adjustments, white balance, automatic and manual exposure, and individual objective calibration system.
Cameras are available in a wide range of resolution and connectivity options for applications ranging from education, conservation, pharmaceutical, or other demanding fields.
The Moticam 4000 is a camera that was designed for ease of use and the ability to output fast live images at 4K(3840x2160) pixels in full HD. Unlike other cameras, the Moticam 4000 can be connected to a computer or through HDMI. Simply plug the camera through the HDMI port and display images with the use of the onboard software and a USB mouse through an HDMI monitor, or connect your camera through USB to a Windows, Mac or Linux computer.
Conformite Europeenne
Specifications
  • Resolution8MP
  • Image Sensor1/1.8" IMX334
  • Data Format(s)Still image JPEG/TIFF; Video MP4
  • System RequirementsMicrosoft Windows 10, Mac OS X, Linux
  • Data TransferUSB and HDMI
  • Power12V
  • DescriptionMicroscope Camera, IMX334 Sensor, 8MP, 3840 x 2160 pixels, USB and HDMI