Motic Instruments Moticam X5 Microscope Camera, CMOS Sensor, 4MP, 1920 x 1080 pixels, Wi-Fi
Representative image only.
Motic Instruments Moticam X5 Microscope Camera, CMOS Sensor, 4MP, 1920 x 1080 pixels, Wi-Fi
Motic Instruments | Mfr: D-MOTICAM X5| Item: 4892598
Capture images and stream video from your Motic® microscopes
₹108,000.00INR / Each
₹108,000.00INR / Each
Product Description
Includes
CS ring adapter, focusable lens, 30mm and 38mm eyepiece adapters, USB 2.0 cable, Motic 4 dot calibration slide, macro tube, and Motic Images Plus 3.0 PC/OSX/Linux.
Key Features
  • Documentation and analysis for a deeper view of your slide
  • Easy to install and ready for immediate use
  • Capture, annotate, measure or evaluate still images and video
  • Moticam X5 features a 1/3" sensor size
  • 1 year warranty
More About this Item
Motic® Moticam microscope cameras are designed for ease of use and the ability to output images and video. These cameras can produce still image and video capture, still image measurement, image adjustments, white balance, automatic and manual exposure, and individual objective calibration system.
Cameras are available in a wide range of resolution and connectivity options for applications ranging from education, conservation, pharmaceutical, or other demanding fields.
The Moticam X5 Plus is a high resolution streaming WiFi camera. By creating its own WiFi signal, simply connect with your Android or iOS device and view, capture, and edit live images from your microscope3
1 Year WarrantyConformite Europeenne
Specifications
  • Resolution4MP
  • Image Sensor1/3" CMOS
  • Data Format(s)Still image JPEG/TIFF; Video MP4
  • System RequirementsWindows XP/Vista/7/8/10 and MAC OSX, Linux, Android 4.0 or higher, iOS 7 or higher
  • Data TransferWi-Fi (2.4 & 5 GHz), Ethernet
  • Power5V
  • DescriptionMicroscope Camera, CMOS Sensor, 4MP, 1920 x 1080 pixels, Wi-Fi
  • Warranty1 year warranty