Motic Instruments Moticam A8 Microscope Camera, sCMOS Sensor, 8MP, 3840 x 2160 pixels, USB
Representative image only.
Motic Instruments Moticam A8 Microscope Camera, sCMOS Sensor, 8MP, 3840 x 2160 pixels, USB
Motic Instruments | Mfr: 1100600101443| Item: 4892594
Capture images and stream video from your Motic® microscopes
₹130,000.00INR / Each
₹130,000.00INR / Each
Product Description
Includes
CS ring adapter, focusable lens, 30mm and 38mm eyepiece adapters, USB 2.0 cable, Motic 4-dot calibration slide, macro tube, and calibration card.
Key Features
  • Documentation and analysis for a deeper view of your slide
  • Easy to install and ready for immediate use
  • Capture, annotate, measure or evaluate still images and video
  • Moticam A8 features a 1/3" sensor size
More About this Item
Motic® Moticam microscope cameras are designed for ease of use and the ability to output images and video. These cameras can produce still image and video capture, still image measurement, image adjustments, white balance, automatic and manual exposure, and individual objective calibration system.
Cameras are available in a wide range of resolution and connectivity options for applications ranging from education, conservation, pharmaceutical, or other demanding fields.
The Moticam A8 uses a fast sCMOS imaging setup to deliver 3840x2160 pixel images at up to 15 frames per second. That’s 4K imaging with 8MP resolution. With its higher resolution, the A8 is also ideal for documentation and flexible image capturing.
Conformite Europeenne
Specifications
  • Resolution8MP
  • Image Sensor1/3" sCMOS
  • Data Format(s)Still image JPEG/TIFF; Video MP4
  • System RequirementsMicrosoft Windows 10, Mac OSX10.9, Linux UBUNTU1604 or higher
  • Data TransferUSB
  • Power5V (USB supply)
  • DescriptionMicroscope Camera, sCMOS Sensor, 8MP, 3840 x 2160 pixels, USB