IKA® RC 2 Series Compact Recirculating Chillers

Compact yet powerful recirculating chillers designed for external cooling applications
  • Large display for easy setting and viewing of parameters
  • RS-232 and USB interfaces—connect to a PC to control the recirculating chillers
Use RC 2 recirculating chillers for fast and efficient cooling of external devices such as rotary evaporators, reflux condensers, and calorimeters. PID temperature control provides excellent accuracy and uniformity. Connect the chiller to a PC via the USB or RS-232 interface; control and monitor with the labworldsoft® laboratory software.
867,000.00 - ₹1,951,000.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature (° C) Refrigerant Description
Availability Pricing
IKA RC 2 lite Recirculating Chiller, –10 to 20°C; 115V WW-28711-70 Mfr # 25007939
COMPARE
–10 R290 RC 2 lite Recirculating Chiller, –10 to 20°C; 115V
₹867,000.00INR / Each
IKA RC 2 basic Recirculating Chiller, –20 to 20°C; 115V WW-28711-71 Mfr # 4171001
COMPARE
–20 R134a RC 2 basic Recirculating Chiller, –20 to 20°C; 115V
₹1,377,000.00INR / Each
IKA RC 2 control Recirculating Chiller with Wireless Controller, –20 to 20°C; 115V WW-28711-72 Mfr # 4173001
COMPARE
–30 R134a RC 2 control Recirculating Chiller with Wireless Controller, –20 to 20°C; 115V
₹1,777,000.00INR / Each
IKA RC 2 GREEN basic Recirculating Chiller, –30 to 20°C; 115V WW-28711-73 Mfr # 25007402
COMPARE
–30 R290 RC 2 GREEN basic Recirculating Chiller, –30 to 20°C; 115V
₹1,498,000.00INR / Each
IKA RC 2 GREEN control Recirculating Chiller with Wireless Controller, –30 to 20°C; 115V WW-28711-74 Mfr # 25007404
COMPARE
–30 R290 RC 2 GREEN control Recirculating Chiller with Wireless Controller, –30 to 20°C; 115V
₹1,951,000.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat