IKA® Benchtop Tube Mill Control Mill Accessories
Eliminate contamination with disposable grinding chamber
  • Easily view and clean with transparent, disposable grinding chamber
  • Durable construction suitable for grinding material
Choose from a wide range of accessories for the IKA Benchtop Tube Mill Control Mills, items 28710-88 and 28710-89. Mills are sold separately.
13,202.00 - ₹75,698.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
IKA MT 40.100 Disposable Grinding Chamber, 40 mL; 100/pk 2871116 Mfr # 0020001173
COMPARE
Polypropylene chamber with stainless steel blade Disposable Grinding Chamber, 40 mL; 100/pk
₹64,840.00INR / Each
IKA MT 40.10 Disposable Grinding Chamber, 40 mL, Sterile; 10/pk 2871117 Mfr # 0020001984
COMPARE
Polypropylene chamber with stainless steel blade Disposable Grinding Chamber, 40 mL, Sterile; 10/pk
₹13,202.00INR / Each
IKA MMT 40.1 Multi-Use Milling Tube; 40 mL 2871119 Mfr # 0020003165
COMPARE
Polyamide chamber, stainless steel blade, polyethylene sealing ring, and POM coupling Multi-Use Milling Tube; 40 mL
₹22,733.00INR / Each
IKA MTT 40.10 Disposable Grinding Chamber with Titanium Beater, 40 mL; 10/pk 2871120 Mfr # 0020015380
COMPARE
Polypropylene chamber with titanium blade Disposable Grinding Chamber with Titanium Beater, 40 mL; 10/pk
₹13,202.00INR / Each
IKA A-MMT 40.100 Abrasion Set with 100 Sealing Rings, 10 Beaters, and 10 Couplings 2871121 Mfr # 0020003378
COMPARE
Stainless steel blade, polyethylene sealing ring, and POM coupling Abrasion Set with 100 Sealing Rings, 10 Beaters, and 10 Couplings
₹15,390.00INR / Each
IKA MT 40P.100 Grinding Chamber with Stainless Steel Beater, 40 mL, Bent 45° Angle; 100/pk 2871123 Mfr # 0020011531
COMPARE
Polypropylene chamber with stainless steel blade Grinding Chamber with Stainless Steel Beater, 40 mL, Bent 45° Angle; 100/pk
₹75,698.00INR / Each