IKA A-MMT 40.100 Abrasion Set with 100 Sealing Rings, 10 Beaters, and 10 Couplings
Representative image only.
IKA A-MMT 40.100 Abrasion Set with 100 Sealing Rings, 10 Beaters, and 10 Couplings
IKA | Mfr: 0020003378| Item: 2871121
Eliminate contamination with disposable grinding chamber
₹15,390.00INR / Each
₹15,390.00INR / Each
Buy It Together
Current Item
IKA A-MMT 40.100 Abrasion Set with 100 Sealing Rings, 10 Beaters, and 10 Couplings
IKA A-MMT 40.100 Abrasion Set with 100 Sealing Rings, 10 Beaters, and 10 Couplings
2871121
₹15,390.00INR / Each
+
IKA 0025002269 Benchtop A 10 Basic Mill; 115 VAC, 50/60 Hz
2871090
₹638,000.00INR / Each
+ =
Total Price 1702390.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easily view and clean with transparent, disposable grinding chamber
  • Durable construction suitable for grinding material
More About this Item
Choose from a wide range of accessories for the IKA Benchtop Tube Mill Control Mills, items 28710-88 and 28710-89. Mills are sold separately.
Specifications
  • MaterialStainless steel blade, polyethylene sealing ring, and POM coupling
  • DescriptionAbrasion Set with 100 Sealing Rings, 10 Beaters, and 10 Couplings