Identi-Plug Plastic Stoppers
Durable and reusable foam stoppers are an economical way to keep the contents of your containers secure. Open-cell foam has a uniform density, leaves no fibers like cotton, is breathable, nonabsorbent, chemcial resistant, heat tolerant, and retains shape after repeated use. Can also be used as a cleaner for pipettes, tubes, vials and precision tubing. Autoclavable.
12,500.00 - ₹28,100.00INR / Pkg of 1000
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Port Size (ID) (mm) Description
Availability Pricing
Indenti-Plug L800-A Plastic Foam Stopper, 6-13mm Openings, 1000/pk WW-06298-70 Mfr # L800-A
COMPARE
Foam 6 to 13 Plastic Foam Stopper, 6-13mm Openings, 1000/pk
₹21,300.00INR / Pkg of 1000
Indenti-Plug L800-B Plastic Foam Stopper, 14-20mm Openings, 1000/pk WW-06298-72 Mfr # L800-B
COMPARE
Foam 14 to 20 Plastic Foam Stopper, 14-20mm Openings, 1000/pk
₹28,100.00INR / Pkg of 1000
Indenti-Plug L800-B2 Plastic Foam Stopper, 21-26mm Openings, 400/pk WW-06298-74 Mfr # L800-B2
COMPARE
Foam 21 to 26 Plastic Foam Stopper, 21-26mm Openings, 400/pk
₹23,000.00INR / Pkg of 400
Indenti-Plug L800-C Plastic Foam Stopper, 27-34mm Openings, 200/pk WW-06298-76 Mfr # L800-C
COMPARE
Foam 27 to 34 Plastic Foam Stopper, 27-34mm Openings, 200/pk
₹18,700.00INR / Pkg of 200
Indenti-Plug L800-D Plastic Foam Stopper, 35-45mm Openings, 100/pk WW-06298-78 Mfr # L800-D
COMPARE
Foam 34 to 45 Plastic Foam Stopper, 35-45mm Openings, 100/pk
₹12,900.00INR / Pkg of 100
Indenti-Plug L800-D2 Plastic Foam Stopper, 46-50mm Openings, 80/PK WW-06298-80 Mfr # L800-D2
COMPARE
Plastic 46 to 50 Plastic Foam Stopper, 46-50mm Openings, 80/pk
₹18,800.00INR / Pkg of 80
Indenti-Plug L800-E Plastic Foam Stopper, 46-65mm Openings, 50/pk WW-06298-82 Mfr # L800-E
COMPARE
Plastic 46 to 65 Plastic Foam Stopper, 46-65mm Openings, 50/pk
₹12,500.00INR / Pkg of 50
Indenti-Plug L800-G Plastic Foam Stopper, 66-75mm Openings, 40/pk WW-06298-84 Mfr # L800-G
COMPARE
Foam 66 to 75 Plastic Foam Stopper, 66-75mm Openings, 40/pk
₹14,400.00INR / Pkg of 40
Showing 1- 8 of 8