Indenti-Plug L800-C Plastic Foam Stopper, 27-34mm Openings, 200/pk
Representative image only.
Indenti-Plug L800-C Plastic Foam Stopper, 27-34mm Openings, 200/pk
Indenti-Plug | Mfr: L800-C| Item: 0629876
₹36,300.00INR / Pkg of 200
₹36,300.00INR / Pkg of 200
Product Description
More About this Item
Durable and reusable foam stoppers are an economical way to keep the contents of your containers secure. Open-cell foam has a uniform density, leaves no fibers like cotton, is breathable, nonabsorbent, chemcial resistant, heat tolerant, and retains shape after repeated use. Can also be used as a cleaner for pipettes, tubes, vials and precision tubing. Autoclavable.
Specifications
  • Height (mm)44.5
  • Port Size (ID) (mm)27 to 34
  • ColorWhite
  • MaterialFoam
  • DescriptionPlastic Foam Stopper, 27-34mm Openings, 200/pk