Indenti-Plug L800-B Plastic Foam Stopper, 14-20mm Openings, 1000/pk
Representative image only.
Indenti-Plug L800-B Plastic Foam Stopper, 14-20mm Openings, 1000/pk
Indenti-Plug | Mfr: L800-B| Item: 0629872
₹54,500.00INR / Pkg of 1000
₹54,500.00INR / Pkg of 1000
Product Description
More About this Item
Durable and reusable foam stoppers are an economical way to keep the contents of your containers secure. Open-cell foam has a uniform density, leaves no fibers like cotton, is breathable, nonabsorbent, chemcial resistant, heat tolerant, and retains shape after repeated use. Can also be used as a cleaner for pipettes, tubes, vials and precision tubing. Autoclavable.
Specifications
  • Height (mm)35
  • Port Size (ID) (mm)14 to 20
  • ColorWhite
  • MaterialFoam
  • DescriptionPlastic Foam Stopper, 14-20mm Openings, 1000/pk