Indenti-Plug L800-D2 Plastic Foam Stopper, 46-50mm Openings, 80/PK
Representative image only.
Indenti-Plug L800-D2 Plastic Foam Stopper, 46-50mm Openings, 80/PK
Indenti-Plug | Mfr: L800-D2| Item: 0629880
₹36,300.00INR / Pkg of 80
₹36,300.00INR / Pkg of 80
Product Description
More About this Item
Durable and reusable foam stoppers are an economical way to keep the contents of your containers secure. Open-cell foam has a uniform density, leaves no fibers like cotton, is breathable, nonabsorbent, chemcial resistant, heat tolerant, and retains shape after repeated use. Can also be used as a cleaner for pipettes, tubes, vials and precision tubing. Autoclavable.
Specifications
  • Height (mm)56
  • Port Size (ID) (mm)46 to 50
  • ColorWhite
  • MaterialPlastic
  • DescriptionPlastic Foam Stopper, 46-50mm Openings, 80/pk