Indenti-Plug L800-B2 Plastic Foam Stopper, 21-26mm Openings, 400/pk
Representative image only.
Indenti-Plug L800-B2 Plastic Foam Stopper, 21-26mm Openings, 400/pk
Indenti-Plug | Mfr: L800-B2| Item: 0629874
₹44,600.00INR / Pkg of 400
₹44,600.00INR / Pkg of 400
Product Description
More About this Item
Durable and reusable foam stoppers are an economical way to keep the contents of your containers secure. Open-cell foam has a uniform density, leaves no fibers like cotton, is breathable, nonabsorbent, chemcial resistant, heat tolerant, and retains shape after repeated use. Can also be used as a cleaner for pipettes, tubes, vials and precision tubing. Autoclavable.
Specifications
  • Height (mm)44.5
  • Port Size (ID) (mm)21 to 26
  • ColorWhite
  • MaterialFoam
  • DescriptionPlastic Foam Stopper, 21-26mm Openings, 400/pk