Environmental Express® Round Wide-Mouth Bottles, HDPE
Won't break like glass
  • Provide easy-access wide-mouth
  • Lightweight design with leakproof cap
Choose between Level 1 or 3 pre-cleaned round wide-mouth bottles. Level 1 bottles are lot controlled, sealed with custody tape, and include a Certificate of Analysis from an independent laboratory. Level 3 bottles meet EPA guidelines, but do not ship with a certificate of analysis. They are ideal for general-use.
4,100.00 - ₹94,500.00INR / Case of 250
29 variations of this product are available.
See All Product Options Below
29 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Level Number Special Features
Availability Pricing
COMPARE
60 1 33 mm cap
₹5,600.00INR / Case of 48
COMPARE
60 3 33 mm cap
₹6,500.00INR / Case of 48
COMPARE
125 1 38 mm cap
₹9,900.00INR / Case of 48
COMPARE
125 3 38 mm cap
₹54,100.00INR / Case of 500
COMPARE
125 3 38 mm cap
₹7,000.00INR / Case of 48
COMPARE
250 1 48 mm cap
₹51,100.00INR / Case of 250
COMPARE
250 1 48 mm cap
₹5,300.00INR / Case of 24
COMPARE
250 3 F217 liner
₹53,700.00INR / Case of 336
COMPARE
250 3 48 mm cap
₹46,600.00INR / Case of 250
COMPARE
500 1 53 mm cap
₹6,700.00INR / Case of 24
COMPARE
500 1 53 mm cap
₹55,100.00INR / Case of 180
COMPARE
500 3 53 mm cap
₹7,800.00INR / Case of 24
COMPARE
500 3 53 mm cap
₹53,200.00INR / Case of 180
COMPARE
1000 1 63 mm cap
₹24,200.00INR / Case of 72
COMPARE
1000 1 63 mm cap
₹5,300.00INR / Case of 12
COMPARE
1000 1 Preserved with 0.5 mL nitric acid, 63 mm cap
₹31,400.00INR / Case of 72
COMPARE
1000 3 63 mm cap
₹5,300.00INR / Case of 12
COMPARE
1000 3 63 mm cap
₹29,700.00INR / Case of 72
COMPARE
2000 1 63 mm cap
₹9,600.00INR / Case of 8
COMPARE
2000 3 63 mm cap
₹7,500.00INR / Case of 8
Showing 1 - 20 of 29