Disposable HDPE Milling Jars
HDPE jar with separate sleeve eliminates cross-contamination
Disposable milling jars eliminate cross-contamination problems. Jars handle loads as small as 75 grams—perfect for research applications. Baffled jars prevent grinding media from slipping and provide highly efficient tumbling action. Made from high-density polyethylene (HDPE), these jars withstand strong chemicals.
Jar sleeves sold separately.
129,000.00 - ₹315,000.00INR / Pkg of 12
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Height (in) Jar Capacity (Gal) Material
Availability Pricing
Disposable HDPE Milling Jars, 0.125 gal (0.5 L); 12/Pk 0417200 Mfr # J-0016
COMPARE
6.6562 0.125 HDPE
₹170,000.00INR / Pkg of 12
Disposable HDPE Milling Jars, 0.25 gal (0.9 L); 6/Pk 0417202 Mfr # J-0032
COMPARE
7.78125 0.25 HDPE
₹129,000.00INR / Pkg of 6
Disposable HDPE Milling Jars, 0.5 gal (1.9 L); 6/Pk 0417204 Mfr # J-0064
COMPARE
9.515625 0.5 HDPE
₹205,000.00INR / Pkg of 6
Disposable HDPE Milling Jars, 1.0 gal (3.8 L); 6/Pk 0417206 Mfr # J-0128
COMPARE
11.3125 1 HDPE
₹284,000.00INR / Pkg of 6
Disposable HDPE Milling Jars, 2.5 gal (9.5 L); 4/Pk 0417208 Mfr # J-0320
COMPARE
12 2.5 HDPE
₹315,000.00INR / Pkg of 4