Corning® Gosselin™ Square Wide-Mouth HDPE Storage Bottles
Lightweight and shatterproof bottles with easy-access wide neck
  • Leakproof and resistant to breakage
  • Perfect for storing liquid, semi liquid, and solid media
  • Square and ergonomic HDPE bottle with a screw cap
  • Measure exact contents with molded graduations (except 150 mL)
  • Sterile and food contact compliant
Square bottles take up less space and are easier to stack and pack into boxes. Wide-neck HDPE bottles offer good chemical resistance, and shatterproof protection for your media. Sterile bottles come fully assembled with screw caps, and placed inside PE bags to avoid cross- contamination. Each case includes a traceability label to retrace each production cycle.
34,400.00 - ₹61,200.00INR / Case of 90
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap Color Material
Availability Pricing
Corning Gosselin Storage Bottle with Screw Cap, 1 L, square, HDPE, sterile; 90/cs 3455170 Mfr # LC1000-13
COMPARE
1000 Red High Density Polyethyene (HDPE)
₹34,400.00INR / Case of 90
Corning Gosselin Storage Bottle with Screw Cap, 2 L, square, HDPE, sterile; 50/cs 3455171 Mfr # LC2000-03
COMPARE
2000 Red High Density Polyethyene (HDPE)
₹56,400.00INR / Case of 50
Corning Gosselin Storage Bottle with Screw Cap, 2.5 L, square, HDPE, sterile; 35/cs 3455172 Mfr # LC2500A-09
COMPARE
2500 White High Density Polyethyene (HDPE)
₹53,200.00INR / Case of 35
Corning Gosselin Storage Bottle with Screw Cap, 150 mL, square, HDPE, sterile; 300/cs 3455173 Mfr # LC150A-05
COMPARE
150 Red High Density Polyethyene (HDPE)
₹54,400.00INR / Case of 300
Corning Gosselin Storage Bottle with Screw Cap, 250 mL, square, HDPE, sterile; 210/cs 3455174 Mfr # LC250-06
COMPARE
250 Red High Density Polyethyene (HDPE)
₹57,400.00INR / Case of 210
Corning Gosselin Storage Bottle with Screw Cap, 500 mL, square, HDPE, sterile; 175/cs 3455175 Mfr # LC500-11
COMPARE
500 Red High Density Polyethyene (HDPE)
₹61,200.00INR / Case of 175