Alconox® Low-Foaming Cleaning Detergents
Biodegradable, concentrated, and free of interfering residue
  • Optimize performance and environmental safety
  • Order new bulk packages for added convenience
These low-foaming cleaning detergents are suitable for use in automatic spray systems. They are ideal for use on glass, metal, plastic, rubber, and other hard materials in your critical cleaning processes. All Alconox cleaners rinse completely with no interfering residue. Alconox® and Liquinox® cleaners are biodegradable. All cleaner are safe, odorless, and FDA compliant.
8,000.00 - ₹71,900.00INR / Case of 4
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Dilution Format Volume (Liters)
Availability Pricing
Alconox Detergent 8 1701 Low-Foaming Ion Free Detergent; 4 x 1 gal. Bottles/Cs WW-17775-10 Mfr # 1701
COMPARE
3:100, 11 pH Liquid 4
Call For Price
Alconox Luminox 1901-1 Low-Foaming Neutral Cleaner; 1 gal. Bottle WW-17775-25 Mfr # 1901-1
COMPARE
3:100, 7 pH Liquid 4
₹29,000.00INR / Each
Alconox Luminox 1901 Low-Foaming Neutral Cleaner; 4 x 1 gal. Bottles/Cs WW-17775-30 Mfr # 1901
COMPARE
3:100, 7 pH Liquid 4
₹70,300.00INR / Case of 4
Alconox Alcojet 1404 Low-Foaming Powdered Detergent; 9 x 4 lb Box/Cs WW-17776-00 Mfr # 1404
COMPARE
1:200, 12 pH Powder 1.8
₹71,900.00INR / Case of 1
Alconox Alcojet 1404-1 Low-Foaming Powdered Detergent; 4 lb Box WW-17776-05 Mfr # 1404-1
COMPARE
1:200, 12 pH Powder 1.8
₹8,000.00INR / Each
Alconox Alcojet 1425 Low-Foaming Powdered Detergent; 25 lb Box WW-17776-50 Mfr # 1425
COMPARE
1:200, 12 pH Powder 11.34
₹47,400.00INR / Carton of 1
Alconox Citrajet 2001-1 Low-Foaming Acid Cleaner; 1 gal. WW-17778-20 Mfr # 2001-1
COMPARE
2:100, 2.5 pH Liquid 4
₹14,500.00INR / Each
Alconox Citrajet 2001 Low-Foaming Acid Cleaner; 4 x 1 gal. Bottles/Cs WW-17778-25 Mfr # 2001
COMPARE
2:100, 2.5 pH Liquid 4
₹57,800.00INR / Case of 4
Showing 1- 8 of 8