Alconox Detergent 8 1701 Low-Foaming Ion Free Detergent; 4 x 1 gal. Bottles/Cs
Representative image only.
Alconox Detergent 8 1701 Low-Foaming Ion Free Detergent; 4 x 1 gal. Bottles/Cs
Alconox | Mfr: 1701| Item: 1777510
Biodegradable, concentrated, and free of interfering residue
Call For Price
Call For Price
Buy It Together
Current Item
Alconox Detergent 8 1701 Low-Foaming Ion Free Detergent; 4 x 1 gal. Bottles/Cs
Alconox Detergent 8 1701 Low-Foaming Ion Free Detergent; 4 x 1 gal. Bottles/Cs
1777510
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Optimize performance and environmental safety
  • Order new bulk packages for added convenience
More About this Item
These low-foaming cleaning detergents are suitable for use in automatic spray systems. They are ideal for use on glass, metal, plastic, rubber, and other hard materials in your critical cleaning processes. All Alconox cleaners rinse completely with no interfering residue. Alconox® and Liquinox® cleaners are biodegradable. All cleaner are safe, odorless, and FDA compliant.
Detergent 8 is a nonionic, detergent for labware washers, PC board washers, power-spray systems, ultrasonic and manual cleaning. Removes oils, resins rosins, and fluxes from PC circuit boards, electronic parts, phosphate sensitive labware, and nuclear contaminated equipment with no interfering conductive residues. Excellent replacement for solvent cleaning.
ALCOJET, ALCONOX, LIQUI-NOX, TERG-A-ZYME-Reg TM Alconox Inc.
Specifications
  • FormatLiquid
  • Volume (gallons)1
  • Volume (Liters)4
  • Dilution3:100, 11 pH
  • DescriptionCleaning Detergent, case of four 1-gallon bottles