Labconco LabSolutions™ Low-Foaming Liquid Detergent; 3.8 L
Representative image only.
Labconco LabSolutions™ Low-Foaming Liquid Detergent; 3.8 L
Labconco | Mfr: 4522000| Item: 9896075
Removes wide range of lab contaminants
Call For Price
Call For Price
Buy It Together
Current Item
Labconco LabSolutions™ Low-Foaming Liquid Detergent; 3.8 L
Labconco LabSolutions™ Low-Foaming Liquid Detergent; 3.8 L
9896075
Call For Price
+ =
Total Price 9300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Rinses completely
  • Temperature stable for use in hot or cold water
  • Phosphate- and chlorine-free
  • Does approximately 180 loads
More About this Item
LabSolutions detergents are specially formulated for Labconco glassware washers.
Low-foaming liquid detergent provides removal of a wide range of contaminants while rinsing off completely. Noncorrosive formulation is safe for use on stainless steel, glass, or plastic. Perfect for use with hot or cold water. Free of phosphates and chlorine. Formulated for Labconco FlaskScrubber and SteamScrubber washers.
Specifications
  • Capacity (Liters)3.8
  • Capacity (Gal)1
  • MaterialLow-foaming liquid
  • Compatible WithAutomatic glassware washer
  • DescriptionLow-Foaming Liquid Detergent