Alconox Citrajet 2001 Low-Foaming Acid Cleaner; 4 x 1 gal. Bottles/Cs
Representative image only.
Alconox Citrajet 2001 Low-Foaming Acid Cleaner; 4 x 1 gal. Bottles/Cs
Alconox | Mfr: 2001| Item: 1777825
Biodegradable, concentrated, and free of interfering residue
₹69,500.00INR / Case of 4
₹69,500.00INR / Case of 4
Buy It Together
Current Item
Alconox Citrajet 2001 Low-Foaming Acid Cleaner; 4 x 1 gal. Bottles/Cs
Alconox Citrajet 2001 Low-Foaming Acid Cleaner; 4 x 1 gal. Bottles/Cs
1777825
₹69,500.00INR / Case of 4
+ =
Total Price 69500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Optimize performance and environmental safety
  • Order new bulk packages for added convenience
More About this Item
These low-foaming cleaning detergents are suitable for use in automatic spray systems. They are ideal for use on glass, metal, plastic, rubber, and other hard materials in your critical cleaning processes. All Alconox cleaners rinse completely with no interfering residue. Alconox® and Liquinox® cleaners are biodegradable. All cleaner are safe, odorless, and FDA compliant.
Citrajet is a low foaming, phosphate-free, concentrated, liquid cleaner, and metal brightener for high peformance removal of metal oxides, scales, salts, and inorganic residues in labware washers, parts washers, power-spray systems, and manual and ultrasonic cleaning. Corrosion inhibited and free rinsing leaving no interfering residues. Ideal for acid washing and rinsing. Dilute 2:100 pH 2.5
ALCOJET, ALCONOX, LIQUI-NOX, TERG-A-ZYME-Reg TM Alconox Inc.
Specifications
  • FormatLiquid
  • Volume (gallons)1
  • Volume (Liters)4
  • Dilution2:100, 2.5 pH
  • DescriptionLiquid Cleaning Detergent; 1 gal, 4 Bottles/Cs