Alconox Alcojet 1425 Low-Foaming Powdered Detergent; 25 lb Box
Representative image only.
Alconox Alcojet 1425 Low-Foaming Powdered Detergent; 25 lb Box
Alconox | Mfr: 1425| Item: 1777650
Biodegradable, concentrated, and free of interfering residue
₹55,400.00INR / Carton of 1
₹55,400.00INR / Carton of 1
Buy It Together
Current Item
Alconox Alcojet 1425 Low-Foaming Powdered Detergent; 25 lb Box
Alconox Alcojet 1425 Low-Foaming Powdered Detergent; 25 lb Box
1777650
₹55,400.00INR / Carton of 1
+ =
Total Price 55400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Optimize performance and environmental safety
  • Order new bulk packages for added convenience
More About this Item
These low-foaming cleaning detergents are suitable for use in automatic spray systems. They are ideal for use on glass, metal, plastic, rubber, and other hard materials in your critical cleaning processes. All Alconox cleaners rinse completely with no interfering residue. Alconox® and Liquinox® cleaners are biodegradable. All cleaner are safe, odorless, and FDA compliant.
Alcojet is nonionic, low foaming powder cleaner for use in labware washers, parts washers, power-spray systems, manual, and ultrasonic cleaning. Corrosion inhibited formula recommended for glassware, metals, plastic, ceramic, porcelain, rubber, and fiberglass. Free rinsing for reliable results and no interfering residues. USDA authorized. Dilute 1:200. pH 11.5.
ALCOJET, ALCONOX, LIQUI-NOX, TERG-A-ZYME-Reg TM Alconox Inc.
Specifications
  • FormatPowder
  • Volume (Liters)11.34
  • Dilution1:200, 12 pH
  • DescriptionPowdered Cleaner; 25 lb Box