Alconox Luminox 1901-1 Low-Foaming Neutral Cleaner; 1 gal. Bottle
Representative image only.
Alconox Luminox 1901-1 Low-Foaming Neutral Cleaner; 1 gal. Bottle
Alconox | Mfr: 1901-1| Item: 1777525
Biodegradable, concentrated, and free of interfering residue
₹21,000.00INR / Each
₹21,000.00INR / Each
Buy It Together
Current Item
Alconox Luminox 1901-1 Low-Foaming Neutral Cleaner; 1 gal. Bottle
Alconox Luminox 1901-1 Low-Foaming Neutral Cleaner; 1 gal. Bottle
1777525
₹21,000.00INR / Each
+ =
Total Price 21000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Optimize performance and environmental safety
  • Order new bulk packages for added convenience
More About this Item
These low-foaming cleaning detergents are suitable for use in automatic spray systems. They are ideal for use on glass, metal, plastic, rubber, and other hard materials in your critical cleaning processes. All Alconox cleaners rinse completely with no interfering residue. Alconox® and Liquinox® cleaners are biodegradable. All cleaner are safe, odorless, and FDA compliant.
Luminox is a low foaming, neutral pH detergent for use in manual, ultrasonic, and machine cleaning. Safe-handling detergent used for non-corrosive cleaning without chelation or alkalinity waste treatment problems. Recommended for residue free cleaning of aluminum and soft metal, medical devices, pharmaceutical equipment, alkaline sensitive labware, and medical instruments in aluminum trays. Free rinsing, eliminating interfering residues. Dilute 3:100. pH 7
ALCOJET, ALCONOX, LIQUI-NOX, TERG-A-ZYME-Reg TM Alconox Inc.
Specifications
  • FormatLiquid
  • Volume (gallons)1
  • Volume (Liters)4
  • Dilution3:100, 7 pH
  • DescriptionCleaning Detergent, one-gallon bottle