YSI 6050000 Proplus Multiparameter Meter, Handheld, Waterproof

Representative image only.
YSI Mfr # 6050000 – Item # WW-59352-00
Modular design allows configuration to your specific application
  • Combine meter, probes, and cable lengths to meet your needs
  • Handheld design offers menu-driven software for added convenience
  • Cables and sensors sold separately.
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.02 pH
  • Buffer RecognitionNIST and USA
  • ISE Resolution0.1 ppm
  • ISE Accuracy±20 mV
  • Conductivity Accuracy±0.5% FS + 1 LSD
  • Min TDS0 to 100 ppt
  • Min Salinity0 to 70 ppt
  • Salinity Accuracy±0.1 ppt
  • Min Dissolved Oxygen0 to 50 ppm
  • Dissolved Oxygen Resolution0.1 or 0.01 ppm
  • Dissolved Oxygen Accuracy±2% FS
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • Temperature compensationAutomatic or manual
  • InterfaceUSB
  • WaterproofYes
  • Battery2 x C batteries (included)
  • Battery life (hr)300
  • Width (cm)8.255
  • Height (cm)21.59
  • Width (in)3 1/4
  • Length (in)2 1/4
  • Height (in)8 1/2
  • DescriptionProplus Multiparameter Meter, Handheld, Waterproof
  • Warranty3 year
3 Year WarrantyConformite EuropeenneIP 67ISO9001:2000 Certified Supplier

Key Features

  • Combine meter, probes, and cable lengths to meet your needs
  • Handheld design offers menu-driven software for added convenience
  • Cables and sensors sold separately.
  • 3 year warranty

More About this Item

The YSI® Professional Plus meter provides plug-and-play capabilities to give you as much functionality as needed. Build your own system by ordering the meter, probe(s), and cables(s) you need without having to purchase additional meters or unnecessary accessories.

Easy-to-use meter features graphic display with detailed messages and instructions that translate to less time reading manuals. Display and keypad are backlit, making them ideal for low-light situations. Meter stores up to 1000 data points with time- and date-stamp for GLP compliance. Meter can be programmed to manually record data or set up as continual logger with user-selectable recording rates. USB connectivity permits software upgrades. Rugged rubberized case protects meter during use. Meter is IP67 waterproof-rated, even without the battery lid.

Measure temperature and your choice of pH, ORP, ammonium, chloride, nitrate, conductivity with automatic salinity compensation, and DO with automatic barometric compensation. Meter accepts either classic polarographic or “instant-on” galvanic DO sensors with screw-on membrane caps. Conductivity measurements are performed with a four-electrode conductivity cell for accurate measurements.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat