YSI Pro30™ Conductivity and Pro2030™ Dissolved Oxygen/Conductivity Meters
Modular meter allows configuration to your specific application
  • Handheld meter offers menu-driven software for added convenience
  • Backlit graphic display and keypad
  • Cables and probes are not included. order separately
The YSI® Professional series provides plug-and-play capabilities to give you a meter with as much functionality as needed. Build your own system by ordering the meter, probe(s), and cable(s) without having to purchase additional meters or unnecessary accessories.
Easy-to-use meters feature graphic display with detailed messages and instructions that translate to less time reading manuals. Rugged rubber over-molded case protects meters during use. Meters are IP67-rated and are waterproof, even without the battery lid.
Conductivity measurements are performed with a four-electrode conductivity cell for accurate measurements over the entire range. The Pro2030™ accepts either classic polarographic or “instant-on” galvanic DO sensors with screw-on membrane caps for ease of maintenance. Two-point DO calibration and automatic barometric compensation provide greater accuracy. It automatically corrects the DO measurement based on measured salinity values.
200,000.00 - ₹259,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Conductivity Meter Type Waterproof
Availability Pricing
YSI Pro30™ Conductivity Meter 5935205 Mfr # 6050030
COMPARE
200 mS/cm Conductivity Yes
₹200,000.00INR / Each
YSI Pro2030™ Dissolved Oxygen/Conductivity Meter 5935214 Mfr # 6052030
COMPARE
200 mS/cm DO/Conductivity Yes
₹259,000.00INR / Each