Thermo Scientific STAR A215 Star A215 pH/Conductivity Benchtop Meter Only

Representative image only.

Thermo Scientific STAR A215 Star A215 pH/Conductivity Benchtop Meter Only

Thermo Scientific | Mfr: STARA2150| Item: 5882536

Durability and dependability make it the ideal choice for every lab, every sample, everyday


Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Buy It Together

Current Item

Thermo Scientific STAR A215 Star A215 pH/Conductivity Benchtop Meter Only
Thermo Scientific STAR A215 Star A215 pH/Conductivity Benchtop Meter Only
5882536

₹269,000.00INR / Each
+ + =
Total Price 507700.00 ADD 3 ITEMS TO CART
Product Description

Key Features

  • Multilanguage interface supports English, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Meets US Pharmacopoeia (USP) requirement
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 2000 data points with time date stamp
  • Individual method passwords provide better security
  • Option of AC power or four AA batteries (sold separtely)
  • 3 year warranty

More About this Item

Thermo Scientific Orion Star A215 meter offers pH, mV, RmV, conductivity, salinity, TDS, resistivity and temperatur. Meter features an intuitive user interface with plain language prompts. A large, backlit graphic LCD shows measurements with temperature along with electrode status, time/date, sample ID, user ID, and calibration data.

GLP-related features include 2000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.

Up to five point automatic pH calibration with USA/NIST and Din buffers, auto-buffer recognition, and recalibration feature to fix calibration errors.

Selectable cell constant for use with 2- or 4- band conductivity probes and selectable reference temperatures of 5, 10, 15, 20, 25°C ensure the most accrate conductivity measurement.

Available as meter only and as complete meter kit configurations.

Meter includes swing-arm electrode stand and universal power adapter.

3 Year WarrantyIP54 RatedISO9001:2000 Certified Supplier
Specifications
  • Meter TypeBenchtop
  • Display typeGraphic LCD
  • ConnectionspH: BNC; Temperature: 8-pin mini-DIN
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.01 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min TDS0 ppt
  • Max TDS200 ppt
  • Min Salinity0 ppt
  • Max Salinity80 ppt
  • Resistivity Resolution2 Ω
  • Resistivity Accuracy±0.5% ±1 digit
  • Dissolved Oxygen Accuracy±0.5% of reading
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • InterfaceRS-232 and USB
  • WaterproofYes
  • Power (VAC)100 to 240
  • Power (Hz)50 or 60
  • Battery4 AA (not included)
  • Width (cm)17.78
  • Height (cm)24.765
  • Width (in)7
  • Length (in)3 3/8
  • Height (in)9 3/4
  • DescriptionStar A215 pH/Conductivity Benchtop Meter Only
  • Warranty3 year
Reviews
Q&A