MYRON L ULTRAPEN Conductivity/TDS/Salinity Testers
Waterproof, pocket-sized meters for water analysis testing
  • Autoranging to select measurement range
  • KCl, NaCl and 442™ natural water modes
  • Automatic temperature compensation
  • Model PTBT1 offers Bluetooth for data transfer
This pen-style pocket tester delivers accurate and reliable measurements of conductivity, TDS, and salinity. It’s easy to use: simply press a button and dip the pen into the solution. The LCD gives you a quick measurement reading, alternating with temperature, available in both a live and hold measurement mode. Autoranging to select measurement range and single-point calibration features for KCl, NaCl and 442 natural water modes. Fully encapsulated electronics and a waterproof body constructed of anodized aircraft aluminum with protective coating provide excellent durability.
49,900.00 - ₹102,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Myron L PT1 ULTRAPEN Conductivity Pen 0575405 Mfr # PT1
COMPARE
MyronL ULTRAPEN Conductivity Tester - +/- 1%
₹49,900.00INR / Each
Myron L PTBT1 ULTRAPEN Conductivity Pen with Bluetooth 0575432 Mfr # PTBT1
COMPARE
MyronL ULTRAPEN Conductivity Pen with Bluetooth
₹102,000.00INR / Each