InnoCal NIST-Traceable Calibration; pH Tester or Pen
Representative image only.
InnoCal NIST-Traceable Calibration; pH Tester or Pen
InnoCal | Item: 1710145
Services that ensure the accuracy of your pH Measurement instrumentation
₹18,700.00INR / Each
₹18,700.00INR / Each
Buy It Together
Current Item
InnoCal NIST-Traceable Calibration; pH Tester or Pen
InnoCal NIST-Traceable Calibration; pH Tester or Pen
1710145
₹18,700.00INR / Each
+
Traceable Calibrated Portable Conductivity Meter
1960103
₹72,600.00INR / Each
+ + + + + + + + =
Total Price 898500.00 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Up to three calibration points against NIST-traceable pH buffers
  • 1 year warranty
More About this Item
Regulatory and quality agencies like FDA and EPA have strict guidelines for the determination and control of pH. Maintain your compliance with the help of InnoCal. We can calibrate new instrument purchases from Cole-Parmer, recalibrate your existing instruments, and provide maintenance and repair services.
InnoCal’s metrologists can service most any brand of pH, conductivity, or dissolved oxygen meter including Oakton Instruments, accumet, Fisher Brand, Mettler-Toledo, YSI, WTW, Horiba, Hanna Instruments, Thermo Scientific Orion, VWR SympHony, and others.
Calibrations are performed on any handheld or benchtop pH meter without a BNC connector. The calibration contains three test points at pH 4, 7 and 10 using standard reference buffers produced by an ISO Guide 34 accredited manufacturer. If present, the automatic temperature compensation (ATC) probe will also be tested.
NIST Traceable Calibration1 Year Warranty
Specifications
  • Calibration TypeLiquid Testing
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points3 pH Points, 1 Temperature Point
  • Calibration Test Points4, 7, 10 pH values, 25 C
  • Calibration Test Range4 to 10 pH
  • DescriptionNIST Traceable Calibration; pH Tester and Pens