Hach HQ2200 Series Portable pH, Conductivity, TDS, Salinity, DO, ORP Meter, 2 Channels
A robust and intuitive multiparameter meter
  • Intellical™ smart probes have “plug-and-play” automatic parameter recognition
  • Easy step-by-step illustrated calibration and troubleshooting procedures reduce errors
  • Store up to 10,000 data points
  • Option to select from 27 languages in setup mode
  • IP67 rating with battery compartment installed
  • GLP-compliant features include time, date, sample ID, operator ID, and calibration
Hach portable HQ meters offer a versatile range of measurements, including pH, conductivity, total dissolved solids (TDS), salinity, dissolved oxygen (DO), oxidation reaction potential (ORP), and temperature. The Intellical™ probes are automatically recognized by the HQ meter and will retain calibration history and method settings to minimize errors and setup time. The proven technology delivers superior accuracy and response time, even when measuring challenging samples or in the field.
The illustrated screen displays step-by-step calibration and troubleshooting procedures on the 536 x 335 Mono-TFT display. Whether you are in the lab or in the field, transfer your measurements in just a few clicks via USB or optional Bluetooth. The optional Hach Communication Dongle (HCD) connects the meter to Claros™ data management software for additional data validation.
372,000.00 - ₹948,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hach HQ2200  Portable Multiparameter pH/Conductivity/TDS/Salinity/DO/ORP, 2 Channel WW-59005-19 Mfr # LEV015.53.2200A
COMPARE
Portable Multi-Meter pH/Conductivity/TDS/Salinity/DO/ORP, 2 Channel
₹372,000.00INR / Each
Hach HQ2200  Portable Multiparameter with  Dissolved Oxygen Electrode, 5 m Rugged Cable WW-59005-20 Mfr # LEV015.53.22008
COMPARE
Portable Multi-Meter with Dissolved Oxygen Electrode, 5 m Rugged Cable
₹727,000.00INR / Each
Hach HQ2200  Portable Multiparameter with General-Purpose pH and Conductivity Electrodes, 1 m Cables WW-59005-21 Mfr # LEV015.53.22001
COMPARE
Portable Multi-Meter with General Purpose pH and Conductivity Electrodes, 1 m Cables
₹707,000.00INR / Each
Hach HQ2200  Portable Multiparameter with Low-Maintenance pH and Conductivity Electrodes, 1 m Cables WW-59005-22 Mfr # LEV015.53.22003
COMPARE
Portable Multi-Meter with Low Maintenance pH and Conductivity Electrodes, 1 m Cables
₹688,000.00INR / Each
Hach HQ2200  Portable Multiparameter with pH and Conductivity Electrodes, 5 m Rugged Cables WW-59005-23 Mfr # LEV015.53.22002
COMPARE
Portable Multi-Meter with pH and Conductivity Electrodes, 5 m Rugged Cables
₹931,000.00INR / Each
Hach HQ2200  Portable Multiparameter with Low-Maintenance pH and Dissolved Oxygen Electrodes, 1 m Cables WW-59005-24 Mfr # LEV015.53.22004
COMPARE
Portable Multi-Meter with Low Maintenance pH and Dissolved Oxygen Electrodes, 1 m Cables
₹765,000.00INR / Each
Hach HQ2200  Portable Multiparameter with General-Purpose pH and Dissolved Oxygen Electrodes, 1 m Cables WW-59005-25 Mfr # LEV015.53.22005
COMPARE
Portable Multi-Meter with General Purpose pH and Dissolved Oxygen Electrodes, 1 m Cables
₹777,000.00INR / Each
Hach HQ2200  Portable Multiparameter with pH and Dissolved Oxygen Electrodes, 5 m Rugged Cables WW-59005-26 Mfr # LEV015.53.22006
COMPARE
Portable Multi-Meter with pH and Dissolved Oxygen Electrodes, 5 m Rugged Cables
₹948,000.00INR / Each
Hach HQ2200  Portable Multiparameter with pH Electrode, 1 m Cable WW-59005-27 Mfr # LEV015.53.22009
COMPARE
Portable Multi-Meter with pH Electrode, 1 m Cable
₹676,000.00INR / Each
Showing 1- 9 of 9