Wildco Ekman Dredge with Case, 6" x 6" x 6" Sample Area
Representative image only.
Wildco Ekman Dredge with Case, 6" x 6" x 6" Sample Area
Wildco | Mfr: 196-B10| Item: 0547000
Quickly sample sediment from various bodies of water
₹200,000.00INR / Each
₹200,000.00INR / Each
Buy It Together
Current Item
Wildco Ekman dredge with case, 6" x 6" x 6" sample area
Wildco Ekman dredge with case, 6" x 6" x 6" sample area
0547000
₹200,000.00INR / Each
+ + =
Total Price 386300.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for collecting samples of bottom fauna for productivity, biological, and geological studies
  • 3 month warranty
More About this Item
Sample soft, silty, vegetation-free river, stream, or lake bottoms. Messenger-activated dredges are constructed of stainless steel with stainless steel springs and cables. Feature flow-through design for low drag and good descent stability. Dredges have positive-acting lid and jaw-control mechanism. Includes a marine plywood carrying case. Order required messenger and descent cable separately.
3 Month Warranty
Specifications
  • MaterialStainless Steel
  • DescriptionEkman dredge with case, 6" x 6" x 6" sample area