Wildco 46-D80 Solid Barrel Messenger, stainless steel, 8 oz

Representative image only.

Wildco 46-D80 Solid Barrel Messenger, stainless steel, 8 oz

Wildco | Mfr: 46-D80| Item: 0549915

Minimize contamination

₹9,900.00INR / Each

₹9,900.00INR / Each

Buy It Together

Current Item

Wildco 46-D80 Solid Barrel Messenger, stainless steel, 8 oz
Wildco 46-D80 Solid Barrel Messenger, stainless steel, 8 oz
0549915

₹9,900.00INR / Each
+ =
Total Price 24000.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Fit up to 1/4" (0.6 cm) line
  • 3 month warranty

More About this Item

Messengers activate closing mechanisms on sampling equipment.

3 Month Warranty
Specifications
  • Sample Volume8 oz
  • DescriptionSolid Barrel Messenger, stainless steel, 8 oz
  • Warranty3 month
Reviews
Q&A