Kemmerer Water Samplers

Ensure closure in fast flowing streams or turbulent waters, regardless of line angle
  • The 304 SS models have urethane end seals (do not use when mercury concentrations exceed 1 ppm)
  • Acrylic models have silicone end seals
Sample at specific depths between 3 and 600 feet. The messenger activates the unique trip heads that ensure closure in fast flowing streams or turbulent waters, regardless of line angle. All models include a plastic carrying case; order messenger and line separately.
171,000.00 - ₹192,000.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Sample Volume
Availability Pricing
Wildco 1204-E30 Kemmerer Water Sampler, 3 to 600 ft Depth, 304 SS, 0.4 L 0548500 Mfr # 1204-E30
COMPARE
Stainless Steel 0.4 L
₹171,000.00INR / Each
Wildco 1200-E30 Kemmerer Water Sampler, 3 to 600 ft Depth, 304 SS, 1.2 L 0548510 Mfr # 1200-E30
COMPARE
Stainless Steel 1.2 L
₹192,000.00INR / Each
Wildco 1510-C20 Kemmerer Water Sampler, 3 to 600 ft Depth, Acrylic, 1.2 L 0548600 Mfr # 1510-C20
COMPARE
Acrylic 1.2 L
₹171,000.00INR / Each
Wildco 1520-C20 Kemmerer Water Sampler, 3 to 600 ft Depth, Acrylic, 2.2 L 0548610 Mfr # 1520-C20
COMPARE
Acrylic 2.2 L
₹183,000.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat