Wildco 45-B10 Split Barrel Messenger, stainless steel, 11 oz

Representative image only.

Wildco 45-B10 Split Barrel Messenger, stainless steel, 11 oz

Wildco | Mfr: 45-B10| Item: 0549910

Minimize contamination

₹15,300.00INR / Each

₹15,300.00INR / Each

Buy It Together

Current Item

Wildco 45-B10 Split Barrel Messenger, stainless steel, 11 oz
Wildco 45-B10 Split Barrel Messenger, stainless steel, 11 oz
0549910

₹15,300.00INR / Each
+ =
Total Price 29400.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Fit up to 1/4" (0.6 cm) line
  • 3 month warranty

More About this Item

Messengers activate closing mechanisms on sampling equipment.

3 Month Warranty
Specifications
  • Sample Volume11 oz
  • DescriptionSplit Barrel Messenger, stainless steel, 11 oz
  • Warranty3 month
Reviews
Q&A