Thermo Scientific Cimarec 50094596 i Poly 15 stirrer 100-240V

Representative image only.

Thermo Scientific Cimarec 50094596 i Poly 15 stirrer 100-240V

Thermo Scientific | Mfr: 50094596| Item: 0465640

Call For Price

for an additional
₹41,400.00INR / Each
for an additional
₹29,300.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Thermo Scientific Cimarec 50094596 i Poly 15 stirrer 100-240V
Thermo Scientific Cimarec 50094596 i Poly 15 stirrer 100-240V
0465640

Call For Price
+ +
InnoCal NIST-Traceable Calibration; Stirrer Only
1705004

₹29,300.00INR / Each
+ + +
SHOWA 1005-Md Latex Gloves, 5 mil, medium
8643151

₹3,800.00INR / Box of 100
+
Whatman 6713-5036 Venting Filter, Capsule, 500 cm2
2970064

₹38,200.00INR / Each
+
Cole-Parmer 40 Pound Nylon Cable Zip Ties, 5.5" L, Black; 100/PK
0683078

₹1,900.00INR / Pkg of 100
+ + =
Total Price 1105000.00 ADD 10 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Memory function stores last speed settings
  • Electromagnetic drive means no moving parts and no maintenance
  • 2 year warranty

More About this Item

Use these Thermo Scientific ® Cimarec stirrers for growing microorganisms, dissolving nutrient mediums and solids, and titration applications. Thermo Scientific Cimarec® Stirrers accommodate a variety of vessel shapes including Erlenmeyer flasks and beakers. Stirring speed remains constant even if viscosity of the medium changes. A three-digit LED indicates stirring speed on the 15-position models. The powder-coated stainless steel housing protects unit from spills and dirt. All stirrers include a 6-ft cord with a power supply.

Note: Not for use in hazardous areas; not immersible.

Two year warranty
Specifications
  • Top Plate Length (in)2 1/2
  • Top Plate Width (in)2 1/2
  • Top Plate Length (cm)6.35
  • Top Plate Width (cm)6.35
  • Top Plate MaterialStainless Steel
  • Low Speed (rpm)130
  • High Speed (rpm)990
  • No. Of Stirring Positions15
  • Max Stirring Volume per position (Liters)0.8
  • Length (in)16 1/2
  • Width (in)9 13/16
  • Height (in)1 5/8
  • Length (cm)41.9
  • Width (cm)24.92
  • Height (cm)4.13
  • Plug TypeUS standard
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Descriptioni Poly 15 stirrer 100-240V
  • Warranty2 year
Reviews
Q&A