InnoCal A2LA ISO17025 Accredited Calibration; Stirrer Only
Representative image only.
InnoCal A2LA ISO17025 Accredited Calibration; Stirrer Only
InnoCal | Item: 1750035
Quick and accurate mixer and stirrer calibrations accredited to ISO/IEC 17025 standards
₹41,400.00INR / Each
₹41,400.00INR / Each
Product Description
Key Features
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification, EPA, USDA, ISO and GLP/GMPs standards
  • A2LA Accredited Calibration. View our scope of accreditation
More About this Item
InnoCal provides calibration services for a wide variety of laboratory and industrial mixing and stirring equipment.
Stirrers and stirring hotplates can also be calibrated for rotational accuracy (rpm). All rotational motion (rpm) measurements are traceable to NIST and include data. Equipment is tested at 10 rpm values throughout the range. Accuracy is confirmed using NIST-traceable optical devices.
Service for stirring hotplates and hotplates adds NIST-traceable, temperature control system calibration.
Major brands of stirrers and stirring hotplates that InnoCal can service are Cole-Parmer, Corning, EchoTherm, Fisher Scientific Isotemp, IKA, Thermo Scientific (Cimarec, Super Nuova), Troemner.
Specifications
  • Calibration TypeA2LA ISO17025 Accredited Mechanical
  • System Calibration (Instrument and Probe)Yes
  • Calibration Test PointsAcross the Range of the Instrument
  • DescriptionA2LA ISO17025 Accredited Calibration; Stirrer Only