Cole-Parmer® MSU-600 Compound Microscopes
Powerful and ergonomic microscopes at an affordable price—ideal for life sciences, biomedical research, and universities
  • Smooth-operating, revolving nosepiece provides effortless objective changes
  • Equipped with a Siedentopf type head with 30° incline for comfort
  • A unique rotating system allows ergonomic positioning of both tubes in a high (431 mm) and in a low (397 mm) position
Easily focus on your sample using coaxial coarse and fine adjustments. Featuring 200 graduations, you can adjust 1 µm per graduation and 200 µm per rotation with total travel of approximately 24 mm. Adjustable rack stop prevents damage to sample and objectives. The coarse adjustment is equipped with friction control.
All models feature LED light for energy efficiency, long life span, and cooler temperature. Built-in sensor automatically turns off illumination shortly after nonuse. Safely transport the microscopes using the integrated carrying handles. Store excess cable in the back of the microscope during use and stow away cable during transport and storage.
326,000.00 - ₹1,037,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Illuminator(s) Objectives
Availability Pricing
COMPARE
Binocular 3 W NeoLED 4x/10x/40x/100x*, E-Plan IOS
₹326,000.00INR / Each
COMPARE
Trinocular 3 W NeoLED 4x/10x/40x/100x*, E-Plan IOS
₹357,000.00INR / Each
COMPARE
Trinocular 3 W Kohler NeoLED 4x/10x/40x/100x*, Plan IOS
₹424,000.00INR / Each
COMPARE
Trinocular 3 W Kohler NeoLED 10x/20x/40x/100x*, Plan Phase IOS
₹634,000.00INR / Each
COMPARE
Trinocular 5 W LED 4x/10x/40x/100x*, Plan IOS
₹719,000.00INR / Each
COMPARE
Trinocular 3 W Kohler NeoLED/Epi-NeoLED illumination 5x/10x/20x/50x*, Plan Metal IOS
₹819,000.00INR / Each
COMPARE
Trinocular 3 W Kohler NeoLED/50 W halogen Epi-illumination 5x/10x/20x/50x*, Plan Polarisation Reflected(illumination) IOS
₹1,037,000.00INR / Each
Showing 1- 7 of 7