Thermo Scientific 9417BN Solid State Half-Cell ISE Electrode; Chloride
Representative image only.
Thermo Scientific 9417BN Solid State Half-Cell ISE Electrode; Chloride
Thermo Scientific | Mfr: 9417BN| Item: 0572141
For a complete system:
₹292,000.00INR / Each
₹292,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific 9417BN Solid State Half-Cell ISE Electrode; Chloride
Thermo Scientific 9417BN Solid State Half-Cell ISE Electrode; Chloride
0572141
₹292,000.00INR / Each
+ + + =
Total Price 588200.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ion-selective electrode
  • Reference electrode
  • Standardizing solution
  • Filling solution for reference electrodes
  • Ionic strength and pH adjustors
  • 1 year warranty
More About this Item
Select the ion-selective electrodes, reference electrodes, and necessary solutions to best fit your application. Choose ion-selective electrodes with BNC connectors and 3-ft cord. Note: All electrodes listed are sensing half-cells which require a reference electrode.
1 Year Warranty
Specifications
  • ApplicationsHalf-cell
  • Electrode TypeSolid-state
  • Reference JunctionSure-Flow
  • Reference Electrolyte Double Junction Sure-Flow Reference Electrode, Inner Chamber Filling Solution
  • Ion-Selective ElectrodeChloride
  • Min pH (pH)2
  • Max pH (pH)12
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • ConnectorBNC
  • DescriptionSolid State Half-Cell ISE Electrode; Chloride