Thermo Scientific 940011 Ionic strength and pH adjustors, ISA 5 M NaNO3
Representative image only.
Thermo Scientific 940011 Ionic strength and pH adjustors, ISA 5 M NaNO3
Thermo Scientific | Mfr: 940011| Item: 0573027
Use these ionic strength and pH adjustors for optimal performance of your ISE
₹25,400.00INR / Each
₹25,400.00INR / Each
Buy It Together
Current Item
Thermo Scientific 940011 Ionic strength and pH adjustors, ISA 5 M NaNO3
Thermo Scientific 940011 Ionic strength and pH adjustors, ISA 5 M NaNO3
0573027
₹25,400.00INR / Each
+ =
Total Price 46900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Calibrate your instrument and help perform addition and subtraction operations
More About this Item
Combine with ionic strength and pH adjusters to eliminate interferences from other ion species and adjust standards and samples to an optimal pH range. Standardizing and filling solutions as well as ion-selective electrodes are sold separately.
Specifications
  • Ion-Selective ElectrodeIodine
  • DescriptionIonic strength and pH adjustors, ISA 5 M NaNO3