Thermo Scientific Reference Outer fill solution

Representative image only.

Thermo Scientific Reference Outer fill solution

Thermo Scientific | Mfr: 900003| Item: 0573040

Have refill solutions handy and minimize downtime

₹27,400.00INR / Carton of 5

₹27,400.00INR / Carton of 5
Product Description

Key Features

  • Five 60 mL bottles
  • 5 x 60mL bottles

More About this Item

Ensure accurate pH readings with reference fill solutions.

Specifications
  • Compatible WithReference electrode 05710-10
  • TypeOuter fill solution
  • DescriptionpH Electrode Reference Outer Filling Solution, 5 x 60 mL
Reviews
Q&A