Thermo Scientific 900200 Sure-Flow Reference ISE Electrode, DJ; Pin
Representative image only.
Thermo Scientific 900200 Sure-Flow Reference ISE Electrode, DJ; Pin
Thermo Scientific | Mfr: 900200| Item: 0571010
Get precision measurements of virtually any sample
₹64,900.00INR / Each
₹64,900.00INR / Each
Buy It Together
Current Item
Thermo Scientific 900200 Sure-Flow Reference ISE Electrode, DJ; Pin
Thermo Scientific 900200 Sure-Flow Reference ISE Electrode, DJ; Pin
0571010
₹64,900.00INR / Each
+
Thermo Scientific 900002 Inner chamber filling solution
0573039
₹27,400.00INR / Carton of 5
=
Total Price 92300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Great for applications where rough handling and breakage are a problem Use in conjunction with a separate reference electrode
  • 3 month warranty
More About this Item
Electrodes combine the Sure-Flow easy-clean junction with the speed, accuracy, and stability of Ross electrodes.
When filled with Orion 05730-39 and 05730-40 fill solutions. The Orion 05710-10 matches the potential characteristics of a conventional KCl saturated calomel electrode, with the added no-clog and stable features of a double-junction probe. Reference electrode Orion 05710-10 is recommended for all pH and ion-selective measurements including:
In dilute solutions where the ion to be measured is contained in the filling solution of the Orion 05710-01
In solutions where an ion in the filling solution would introduce an electrode interference.
3 Month Warranty
Specifications
  • ApplicationsHalf cell, reference
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionSure-flow
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)0
  • Max pH (pH)14
  • ConnectorPin-tip (U.S. standard)
  • Diameter (mm)13
  • Length (mm)110
  • Cable Length (m)1
  • DescriptionSure-Flow Reference ISE Electrode, DJ; Pin